Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic Acid, 85%, FCC, 81-89%, Spectrum™

Click to view available options
Quantity:
125 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
50-21-5
C3H6O3
90.08
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
2-hydroxypropanoic acid
CC(O)C(O)=O
50-21-5
C3H6O3
90.08
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
2-hydroxypropanoic acid
CC(O)C(O)=O

L1009, 50-21-5, C3H6O3

Spectrum™ Lactic Acid, 85 Percent, FCC is used as a food preservative, curing agent, and flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Chloride 0.001
UN Number UN3265
Percent Purity 81 to 89%
Ignition Residue 0.001
Lead (Pb) 0.5 mg/kg
Packaging Poly Bottle
Sulfate 0.0025
Quantity 6 x 500 mL