Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactobionic Acid, EP, 98-102%, Spectrum™

Click to view available options
Quantity:
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail
96-82-2
C12H22O12
358.30
JYTUSYBCFIZPBE-AMTLMPIISA-N
(2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanoic acid
OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(O)=O
96-82-2
C12H22O12
358.30
JYTUSYBCFIZPBE-AMTLMPIISA-N
(2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanoic acid
OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(O)=O

L1604, 96-82-2, C12H22O12

Spectrum™ Lactobionic Acid, EP is also known as Galactosylgluconic acid. It is a food additive used as a stabilizer and in the pharmaceutical industry as an excipient for formulation. The EP grade indicates it is grade suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Water 0.05
Quantity 500 g
Specific Rotation +23.0° to +29.0°
CAS Max % 1
Percent Purity 98 to 102%