Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferricin Antibody (5F12.1.2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311195
Description
Lactoferricin Monoclonal specifically detects Lactoferricin in Bovine samples. It is validated for Western Blot, Immunoassay.Specifications
Lactoferricin | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunoassay | |
Bovine Lactoferricin, Lactoferricin B, LF, LF-B, Lfcin, Lfcin B | |
Monoclonal antibody clone 5F12.1.2, anti bovine Lactoferricin B is highly specific for bovine Lactoferricin B. | |
50 μg | |
Primary | |
Bovine | |
Purified |
Western Blot, Immunoassay | |
5F12.1.2 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
4057 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction