Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Antibody (006), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289794JF646
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
| Lactoferrin | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 6 | |
| ELISA, Sandwich ELISA Detection | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Lactoferrin (P02788; Met 1-Lys 710). | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules | |
| 4057 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction