Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lactoferrin Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
$482.50 - $1219.00
Specifications
Assay Range | 78.125 pg/mL |
---|---|
Assay Sensitivity | 78.125-5000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Lactoferrin.Specifications
78.125 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
LTF | |
Amino Acids Drugs and other small molecules | |
Lactoferrin |
78.125-5000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Lactoferrin. | |
Sandwich ELISA | |
4057 | |
Rabbit | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title