Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Mouse anti-Human, DyLight 488, Clone: LF5-1D2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267260G
Description
Lactoferrin Monoclonal antibody specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Lactoferrin | |
LF5-1D2 | |
DyLight 488 | |
Purified | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
LTF | |
Human lactoferrin | |
Monoclonal | |
>95% by SDS-PAGE | |
Amino Acids Drugs and other small molecules | |
Human |
Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen) | |
1mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular) | |
50MM Sodium Borate with 0.05% Sodium Azide | |
4057 | |
Mouse | |
IgG1 | |
Primary | |
0.1mL | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title