Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lactoperoxidase (Bovine)

Used for Enzyme

Manufacturer:  Abnova™ P5202

Catalog No. 89-950-328


Add to cart

Description

Description

#REF!
Specifications

Specifications

Native protein purified from Bovine Milk
Lyophilized
Lactoperoxidase (Bovine)
10 mg
RUO
Lactoperoxidase Assay
Bovine
Enzyme Activity
77.5kDa
Native protein purified from Bovine Milk
Store at -20°C.
LACTOPEROXIDASE
≥35 units per mg dry weight. One Unit reduces one micromole of hydrogen peroxide per minute at 25°C, pH 6.0.
None
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit