Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactose, Monohydrate, Powder, N.F., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

64044-51-5
C12H24O12
360.312
WSVLPVUVIUVCRA-ZEAJSSQNSA-N
133126686
(3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
Packaging Poly Bottle
Formula Weight 360.32
Physical Form Powder
Quantity 2.5kg
Specific Gravity 1L = 1.53kg
Chemical Name or Material Lactose, Monohydrate
Grade NF