Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen LAD1
Concentration 0.31 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-0745
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
161361AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

Immunohistochemical staining of 16136-1-AP shows strong extracellular matrixpositivity in human testis. The predicted MW of this protein is 57 kDa. IP results of 16136-1-AP detected a 62 kDa band.

Anchoring filament protein which is a component of the basement membrane zone.
Specifications

Specifications

LAD1
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Human, Mouse
IgG
-20°C
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit