Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina Biological™ Lady Beetle 10-Larvae Culture Products for Science Education

Observe all 4 stages of complete metamorphosis with lady beetle larvae.

$18.25 - $26.50

Specifications

Animal Yes
Cell Type No
For Use With (Application) CarolinaTM lady bug larvae are an excellent alternative to butterflies in examining insect metamorphosis
Quantity 10 to 15
Source USA
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
S72461 Products for Science Education
View Documents
Carolina Biological™
144218LC
Coupon
Each for $26.50
Add to cart
 
S72460 Products for Science Education
View Documents
Carolina Biological™
144218
Live Specimen
Each for $18.25
Add to cart
 
Description

Description

  • Alligator-like larvae grow, pupate and metamorphose in 10 to 14 days.
  • At least 5 of the 10 to 15 larvae guaranteed to emerge as healthy adult lady beetles.

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

Specifications

Yes
CarolinaTM lady bug larvae are an excellent alternative to butterflies in examining insect metamorphosis
USA
No
10 to 15
22 to 25°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit