Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laemmli SDS Sample Buffer with Pyronin Y, non-reducing, 4X

For protein sample preparation to be used in the Laemmli SDS-PAGE system

$29.70 - $48.85

Specifications

Concentration 4X
Format Liquid
Color Pink
For Use With (Application) For protein sample preparation to be used in the Laemmli SDS-PAGE system
Product Type Sample Loading Buffer
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ61716AC View Documents Alfa Aesar™
J61716AC
25 mL Each for $29.70
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ61716AD View Documents Alfa Aesar™
J61716AD
50 mL Each for $48.85
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

4X
Pink
Sample Loading Buffer
Liquid
For protein sample preparation to be used in the Laemmli SDS-PAGE system
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit