Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laforin/EPM2A Mouse anti-Human, Biotin, Clone: S84-37, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242195B

Catalog No. NBP242195B


Add to cart

Description

Description

Laforin/EPM2A Monoclonal antibody specifically detects Laforin/EPM2A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Laforin/EPM2A
S84-37
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
EPM2A
Fusion protein amino acids 1-331 (full-length) of human Laforin. Rat: 90% identity (296/327 amino acids identical). Mouse: 89% identity (295/327 amino acids identical)
Protein G purified
Protein Phosphatase
Primary
7957
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
EC 3.1.3.16, EC 3.1.3.48, epilepsy, progressive myoclonus type 2, Lafora disease (laforin), epilepsy, progressive myoclonus type 2A, Lafora disease (laforin), EPM2, Lafora PTPase, laforin, LAFPTPase, LD, LDE, MELF
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 40kDa.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit