Learn More
Laforin/EPM2A Mouse anti-Human, DyLight 755, Clone: S84-37, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242195IR
Catalog No. NBP242195IR
Add to cart
Description
Description
Laforin/EPM2A Monoclonal antibody specifically detects Laforin/EPM2A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Laforin/EPM2A
S84-37
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
EPM2A
Fusion protein amino acids 1-331 (full-length) of human Laforin. Rat: 90% identity (296/327 amino acids identical). Mouse: 89% identity (295/327 amino acids identical)
Protein G purified
Protein Phosphatase
Primary
7957
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.1.3.16, EC 3.1.3.48, epilepsy, progressive myoclonus type 2, Lafora disease (laforin), epilepsy, progressive myoclonus type 2A, Lafora disease (laforin), EPM2, Lafora PTPase, laforin, LAFPTPase, LD, LDE, MELF
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 40kDa.