Learn More
LAG-3 Mouse anti-Human, Alexa Fluor™ 750, Clone: 1009614, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB23194S100UG
Catalog No. FB23194S100
Add to cart
This item is not returnable. View return policy
Description
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
LAG-3
1009614
Flow Cytometry 0.25-1 μg/106 cells
P18627
CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC
Mouse myeloma cell line NS0-derived recombinant human LAG-3, Met1-Leu450, Accession # P18627
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human LAG-3 in direct ELISAs.
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Conjugated, Purified, Supernatant
Mouse
IgG2a
100 μg
Primary
3902
Human