Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 Mouse anti-Human, Clone: 874512, R&D Systems™
Mouse Monoclonal Antibody
$123.33 - $385.56
Specifications
Antigen | LAG-3 |
---|---|
Clone | 874512 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB23196100
|
R&D Systems
MAB23196100 |
100 μg |
Each for $385.56
|
|
MAB23196SP
|
R&D Systems
MAB23196SP |
25 μg |
Each for $123.33
|
|
Description
LAG-3 Monoclonal specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
LAG-3 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
Mouse myeloma cell line NS0-derived recombinant human LAG-3, Leu23-Leu450, Accession # P18627 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
874512 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P18627 | |
3902 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title