Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI8F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271112JF646

Catalog No. N271112J646


Add to cart

Description

Description

LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

LAG-3
OTI8F6
Immunohistochemistry
Affinity Purified, Conjugated
LAG3
Recombinant protein fragment corresponding to amino acids 66-332 of human LAG-3 (NP_002277) produced in E.coli.
Immunogen affinity purified
Adaptive Immunity, Immunology, Innate Immunity
Primary
3902.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD223, CD223 antigen, FDC, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit