Learn More
Sklar InstrumentsSupplier Diversity Partner Metzenbaum-Lahey Dissecting Scissors, Sklar, 6 in., Blunt/Blunt, Curved, Smooth, Delicate, Finger Rings, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 221057
Metzenbaum-Lahey Dissecting Scissors, Sklar, 6 in., Blunt/Blunt, Curved, Smooth, Delicate, Finger Rings, Stainless Steel. Sklar Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. This product has curved, smooth blades, blunt tips, and an overall length of 6 inches. This instrument also has a delicate pattern.
Catalog No. 50-143-7850