Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR1 Mouse anti-Human, Janelia Fluor 549, Clone: lc12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104317JF549

Catalog No. NBP14317549


Add to cart

Description

Description

LAIR1 Monoclonal antibody specifically detects LAIR1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

LAIR1
lc12
Flow Cytometry
Purified
LAIR1
Recombinant human LAIR-1 protein
Protein G purified
Store at 4°C in the dark.
Monoclonal
Anti-Hu LAIR-1 mAb, clone lc12, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant extracellular domain(22-125aa) of LAIR-1.
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1
Mouse
IgG1
0.1mL
Primary
3903
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit