Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAIR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121802
This item is not returnable.
View return policy
Description
LAIR2 Polyclonal antibody specifically detects LAIR2 in Human samples. It is validated for Western Blot.Specifications
LAIR2 | |
Polyclonal | |
Unconjugated | |
LAIR2 | |
CD306; CD306 antigen; LAIR2; LAIR-2; leukocyte associated immunoglobulin like receptor 2; leukocyte-associated Ig-like receptor 2; leukocyte-associated Ig-like receptor-2; leukocyte-associated immunoglobulin-like receptor 2; MGC71634 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3904 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q6ISS4 | |
LAIR2 | |
Thr21-Pro152 (Accession Q6ISS4), with N-terminal His Tag | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction