Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAIR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Specifications
Antigen | LAIR2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121802
![]() |
Thermo Scientific
PA5121802 |
100 μL |
Each for $405.00
|
N/A |
PIPA5121803
![]() |
Thermo Scientific
PA5121803 |
20 μL |
Each for $143.00
|
N/A |
Description
LAIR2 Polyclonal antibody specifically detects LAIR2 in Human samples. It is validated for Western BlotSpecifications
LAIR2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
LAIR2 | |
Q6ISS4 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
LAIR2 | |
Primary | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title