Learn More
Lakeland Industries Hi-Vis Public Safety Vests: Polyester
Highly visible vests designed for emergency services, public safety and traffic control personnel
Manufacturer:  Lakeland Industries V+AFOSP2GBVLO
 View more versions of this product
Catalog No. 19-813-359
Add to cart
Description
Description
Sized to fit over operational clothing such as structural fire gear. Appropriate for all types of light and weather conditions.
  • Dual certified—ANSI™ 107-2010, Class 2 and ANSI 207-2006
  • Constructed of FR treated solid polyester or mesh polyester
  • Five-point breakaway design: hook-and-loop front, shoulders and adjustable sides
  • Radio pocket with trim, two mic tabs, inside pocket and badge tab
  • Gro Grain™ trim
  • Color: Lime/Yellow
Compliance
Meet ANSI 107-2010 Class 2 and ANSI 207-2006 and Federal legislation 23CFR634
Specifications
Specifications
Hook and loop closure
Lime
3X- to 5X-Large
2.0
Polyester