Lakeland Industries MicroMax™ Coveralls
Manufacturer: Lakeland Industries TG412LG
 View more versions of this product
Catalog No. 19-810-502
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM D3776, ASTM D5034, ASTM D5034, ASTM D5733, ASTM D5733
    Material Microporous Film Laminate
    Size Large
    Disposable Yes
    For Use With (Application) Protects against dirt, grime, splashes, and spills, ideal for warmer work environments
    Closure Type Zipper front
    Waist Style Elastic
  • Description & Specifications
Related Products
Product Content Feedback