Learn More
Lakeland Industries Nylon Level A Training Suit
$161.00 - $169.00
Specifications
Closure Type | Storm flap |
---|---|
Description | Encapsulated suit with expanded back, PVC faceshield, single storm flap, Butyl gloves, 2 exhaust ports and attached sock boots; PVC Faceshield, Butyl Gloves |
For Use With (Application) | Ideally used as protective apparel from hazardous material |
Material | Nylon |
Ankle Style | Attached boots |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-146-291
|
Lakeland Industries
95493LG |
Large |
Each for $161.00
|
|
19-146-292
|
Lakeland Industries
95493XL |
X-Large |
Each for $161.00
|
|
19-146-293
|
Lakeland Industries
954932X |
2X-Large |
Each for $169.00
|
|
Specifications
Storm flap | |
Ideally used as protective apparel from hazardous material | |
Attached boots |
Encapsulated suit with expanded back, PVC faceshield, single storm flap, Butyl gloves, 2 exhaust ports and attached sock boots; PVC Faceshield, Butyl Gloves | |
Nylon | |
Stitched |