Lakeland Industries Nylon Level A Training Suit
Manufacturer: Lakeland Industries 95493LG
 View more versions of this product
Catalog No. 19-146-291
  Check Availability
Add to Cart
    Specifications
    Material Nylon
    Includes Encapsulated suit with expanded back, PVC faceshield, single storm flap, Butyl gloves, 2 exhaust ports and attached sock boots
    Seam Type Sewn
    Size Large
    For Use With (Application) Ideally used as protective apparel from hazardous material
    Closure Type Single Storm Flap
    Construction PVC Faceshield, Butyl Gloves
  • Description & Specifications
Related Products
Product Content Feedback