Learn More
Lakeland Industries Polypropylene Coveralls With Elastic Wrists/Ankles
Manufacturer:  Lakeland Industries C24174X40
 View more versions of this product
Catalog No. 19-059-815
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Zipper Front
Zipper front, elastic wrist and ankle
White
Yes
Spunbond Polypropylene
Serged
4X-Large
Ideal to protect against dry particulates as well as spray and splatter from water and other non-oil-based liquids