Lakeland Industries Polypropylene Coveralls With Elastic Wrists/Ankles

Manufacturer: Lakeland Industries C24173X40

 View more versions of this product

Catalog No. 19-059-801

  Check Availability
Add to Cart

    Specifications

    Material Spunbond Polypropylene
    Includes Zipper front, elastic wrist and ankle
    Seam Type Serged
    Color White
    Size 3X-Large
    Disposable Yes
    For Use With (Application) Ideal to protect against dry particulates as well as spray and splatter from water and other non-oil-based liquids
    Closure Type Zipper Front

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit