Learn More
Lakeland Industries Level A Test Kit
Fits Lakeland Level A Interceptor suits.
Manufacturer:  Lakeland Industries PTK10
Catalog No. 19-813-262
Add to cart
Requires : Air compressor
Description
Description
  • Features Quick connect fittings for suit air valves, clear air tubing, air supply quick disconnect and timer
Specifications
Specifications
Magnehelic pressure gauge, digital timer, brass and steel fittings, hoses and connectors; portable waterproof case
Tychem Level A encapsulated suit