Learn More
Lakeland Industries Pyrolon Plus 2 Coveralls with Attached Hood
Manufacturer:  Lakeland Industries 07428MD
 View more versions of this product
Catalog No. 19-088-2345
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 2113, 201, ANSI ISEA 101
Elastic
Attached hood
Medium
Zipper
Fabric
Serged
Yes