Learn More
Lakeland Industries Pyrolon Plus 2 Coveralls with Collar
Click to view available options
Size:
Large
2X-Large
3X-Large
Color:
Blue
White
Description:
With collar and zipper
With collar, zipper, elastic wrist and ankle
This item is not returnable. View return policy
This item is not returnable. View return policy
Size 2X-Large
Material Fabric
Color White
Disposable Yes
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Serged
Certifications/Compliance NFPA 2113, 201, ANSI ISEA 101
Description With collar, zipper, elastic wrist and ankle