Lakeland Industries Tychem™ SL Coveralls

Manufacturer: Lakeland Industries 44414LG

 View more versions of this product

Catalog No. 19-088-1935C

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Certifications/Compliance ASTM D1117-80, ASTM D1777-64, ASTM D53034-90, ASTM D3776-85, ASTM D3786-87, ASTM F1001
    Color White
    Material Tyvek™, Saranex™ 23-P Film
    Includes Zipper with storm flap, attached hood, boots, elastic wrist
    Seam Type Sewn and bound
    Size Large
    Disposable Yes
    For Use With (Application) Protection against a broad range of hazardous liquids, toxic chemicals and minute particles
    Resistance Hazardous liquids, toxic chemical and minute particles
    Closure Type Zipper with Storm Flap
    Construction Tyvek™ and laminated with Saranex™ 23-P film

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit