Learn More
Lakeland Industries Polypropylene Coveralls With Zipper
Lightweight, breathable comfort and protection
Manufacturer:  Lakeland Industries C24122XN40
 View more versions of this product
Catalog No. 19-059-363E
Add to cart
Description
Description
  • Serged seams
  • Protects against dry particulates
  • Breathable, porous material helps maintain temperature and comfort
  • Disposable
  • 25 coveralls per case
  • Available in white or navy
Specifications
Specifications
Open
Zipper
Open
2X-Large
4.08kg
Safety
Navy
Polypropylene
9 lb.