Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lakeland Industries ZoneGard Polypropylene Coveralls with Zipper and Collar

For protection in environments where hazardous or nonhazardous materials are present

Manufacturer:  Lakeland Industries C2412SM40

 View more versions of this product

Catalog No. 19-054125


Add to cart

Description

Description

  • Lightweight and breathable
  • Serged seams keep out dry particulates while material remains cool
  • Breathable, porous polypropylene material helps maintain temperature and comfort
  • Color: white
  • Disposable
  • 25 coveralls per case

Specifications

Specifications

Safety
White
Non-toxic workplace
152 - 171cm
Hooded
0.078 lb.
Zipper
Yes
5 ft. - 5 ft. 7 in.
Polypropylene
Small
0.035kg
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit