Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lakeland Industries ZoneGard Polypropylene Coveralls with Zipper and Collar

For protection in environments where hazardous or nonhazardous materials are present

$103.90 - $112.70

Specifications

Closure Type Zipper
Color White
Material Polypropylene
Disposable Yes
For Use With (Application) Non-toxic workplace
View More Specs

Products 2
Catalog Number Mfr. No. Size Height (English) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Height (English) Height (Metric) Price Quantity & Availability  
19-054125
View Documents
Lakeland Industries
C2412SM40
Small 5 ft. 0 in. to 5 ft. 7 in. 152 to 171 cm
Case of 25 for $103.90
Add to cart
 
19-813-298
View Documents
Lakeland Industries
024122X
2X-Large 5 ft. 11 in. to 6 ft. 4 in. 180 to 193 cm
Case of 25 for $112.70
Add to cart
 
Description

Description

  • Lightweight and breathable
  • Serged seams keep out dry particulates while material remains cool
  • Breathable, porous polypropylene material helps maintain temperature and comfort
  • Color: white
  • Disposable
  • 25 coveralls per case

Specifications

Specifications

Zipper
Polypropylene
Non-toxic workplace
0.035 kg
White
Yes
0.078 lb.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit