Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Lalistat 1

Click to view available options
Quantity:
10 mg
50 mg
Description
Lalistat 1 is a potent and selective lysosomal acid lipase (LAL) inhibitor (IC50 = 68 nM). Exhibits no significant activity against pancreatic lipase or lipoprotein lipase at 10 μM concentration. Blocks LAL-mediated lipid hydrolysis of acetylated LDL and reduces efflux of cholesterol from lipid-loaded cells resulting in increased cellular cholesterol ester levels.
Specifications
Specifications
Chemical Name or Material | 4-(Piperidin-1-yl)-1,2,5-thiadiazol-3-yl morpholine-4-carboxylate |
CAS | 501104-16-1 |
Assay Percent Range | >98% |
Quantity | 50 mg |
Molecular Formula | C12H18N4O3S |
Formula Weight | Observed MW: 298.36 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction