Learn More
Thermo Scientific™ Lambda DNA/HindIII Marker Available on GSA/VA Contract for Federal Government customers only.
Perform sizing and approximate quantification of large double-stranded DNA fragments in agarose gels with this conventional marker composed of 8 sharp bands.
Manufacturer:  Thermo Scientific™ SM0101
 View more versions of this product
Catalog No. FERSM0101
Add to cart
Description
Description
Lambda DNA/HindIII Marker is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientitfic restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
The marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively, and it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.
Highlights
  • Sizing and approximate quantification of large DNA fragments
  • Sharp bands
  • Supplied with loading dye for sample DNA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
6X DNA Loading Dye, 2 x 1mL
5 × 50μg