Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Lambda Light Chain Ab-1, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Lambda Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$145.00 - $480.00

Specifications

Antigen Lambda Light Chain Ab-1
Clone HP6054
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS180P
View Documents
Thermo Scientific™
MS180P
Purified Ab with BSA and Azide 1mL Each for $480.00
Add to cart
 
MS180P0
View Documents
Thermo Scientific™
MS180P0
Purified Ab with BSA and Azide 100μL Each for $145.00
Add to cart
 
MS180P1ABX
View Documents
Thermo Scientific™
MS180P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $312.00
Add to cart
 
MS180PABX
View Documents
Thermo Scientific™
MS180PABX
Ab without BSA/Azide (Frozen) 200μL Each for $404.00
Add to cart
 
MS180R7
View Documents
Thermo Scientific™
MS180R7
Ready-to-use for IHC (UVLP) 7mL Each for $279.50
Add to cart
 
MS180P1
View Documents
Thermo Scientific™
MS180P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS180R1
View Documents
Thermo Scientific™
MS180R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Lambda Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: HP6054

Isotype: IgG2a/k

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Lambda Light Chain Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2a kappa
HP6054
Monoclonal
Murine
Hematopoietic
Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit