Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Lambda Light Chain Ab-4 (Cocktail) Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia™ MS1158R1

 View more versions of this product

Catalog No. MS1158R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: LAM03 + HP6054

Isotype: IgG1/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-4 cocktail is especially designed for sensitive detection of lambda light chain of immunoglobulin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Lambda Light Chain Ab-4 (Cocktail)
Monoclonal
Unconjugated
Mouse
1 mL
Hematopoietic
Human
IgG2a κ
Immunohistochemistry (Paraffin)
LAM03 + HP6054
Ready-to-use for IHC (UVLP)
Purified human lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit