Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda Light Chain Mouse anti-Human, Allophycocyanin/Cy7, Clone: 4C2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP145071APCCY7
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
Lambda Light Chain | |
4C2 | |
Flow Cytometry | |
Conjugated, Purified | |
IGLC1 | |
The antibody 4C2 reacts with lambda light chains (22.5 kDa) of human immunoglobulin. | |
Protein A purified | |
Adaptive Immunity, Immunology | |
Primary | |
3537.0 | |
Human |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
IGLC, immunoglobulin lambda constant group | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
The mouse monoclonal antibody H4B4 recognizes CD107b/ LAMP-2, an extensively glycosylated 100-120 kDa widely expressed lysosome-associated protein. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title