Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda Light Chain Mouse anti-Human, Clone: Mc24-IC6, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Lambda Light Chain |
---|---|
Clone | Mc24-IC6 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:10-1:20, ELISA 1:100-1:2000 |
Applications | Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10063014
|
Novus Biologicals
NB10063014 |
0.1 mg |
Each for $436.00
|
|
NB017530
|
Novus Biologicals
NB100630140.025MG |
0.025 mg |
Each for $176.85
|
|
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
Lambda Light Chain | |
0.5 mg/mL | |
Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Immunology | |
P0CG04 | |
3536 | |
Purified human IgG. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mc24-IC6 | |
Flow Cytometry 1:10-1:20, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IGLC, immunoglobulin lambda constant group | |
IGLC1 | |
IgG1 | |
Protein A purified | |
Recognises human lambda light chains, and shows no cross- reactivity to kappa light chains or to immunoglobulin heavy chains. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title