Learn More
Lambda Light Chain Mouse anti-Human, DyLight 650, Clone: 1-155-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179124C
Catalog No. NBP179124C
Add to cart
Description
Description
Lambda Light Chain Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Lambda Light Chain
1-155-2
Flow Cytometry
Purified
IGLC1
Human IgA1 lambda myeloma protein
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody 1-155-2 recognizes lambda light chains (22.5 kDa) of human immunoglobulin.
Flow Cytometry
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
IGLC, immunoglobulin lambda constant group
Mouse
IgG1 Kappa
0.1 ml
Adaptive Immunity, Immunology
Primary
3536
Human