Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Mouse anti-Human, PE-Cyanine7, Clone: ICO-106, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP233308PECY7

Catalog No. N233308PEC7


Add to cart

Description

Description

Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Lambda Light Chain
ICO-106
Flow Cytometry
Conjugated, Purified
IGLC1
Human IgG was used as immunogen to generate the Lambda antibody.
Protein G purified
Adaptive Immunity, Immunology
Primary
3537.0
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
IGLC, immunoglobulin lambda constant group
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
This MAb is specific to lambda light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the lambda light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit