Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Mouse anti-Human, PerCP, Clone: 1-155-2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP179124PCP

Catalog No. NBP179124JP


Add to cart

Description

Description

Lambda Light Chain Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Lambda Light Chain
1-155-2
Flow Cytometry
Purified
IGLC1
Human IgA1 lambda myeloma protein
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody 1-155-2 recognizes lambda light chains (22.5 kDa) of human immunoglobulin.
Flow Cytometry
PerCP
PBS with 0.05% Sodium Azide
IGLC, immunoglobulin lambda constant group
Mouse
IgG1 Kappa
0.1 ml
Adaptive Immunity, Immunology
Primary
3536
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit