Learn More
Lambda Monoclonal Antibody (LcN-2), Zeta
Mouse Monoclonal Antibody Cocktail
Supplier: ZETA CORPORATION Z2457MS
Description
A recommended positive control tissue for this product is Tonsil, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
Lambda Monoclonal antibody specifically detects Lambda in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Lambda | |
Monoclonal | |
200 μg/mL | |
tris with BSA, NP-40 and <0.1% sodium azide | |
P0CG04 | |
Mouse | |
Protein A | |
RUO | |
3535, 3537, 3546 | |
2-8°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
LcN-2 | |
Unconjugated | |
IGLC1 | |
Bence Jones Protein; BJP; Clambda1; Ig lambda chain V-I region BL2; IgL; Igl@; IGLC; IGLC 1/2/3; IGLC1; Igl-C1; IGLL5; IGLV; IGLV@; immmunoglobulin lambda light chain variable region; Immunoglobulin L; immunoglobulin lambda chain complex; immunoglobulin lambda constant 1; immunoglobulin lambda constant 7; immunoglobulin lambda light chain; immunoglobulin lambda light chain constant region; immunoglobulin lambda light chain V-J region; immunoglobulin lambda variable cluster; Lambda LC; lambda-immunoglobulin; LOC100060608; Mcg Marker; Paraprotein | |
Purified human IgG | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.