Learn More
Aviva Systems Biology Corp LAMC1 Antibody - middle region (ARP58234_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58234_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAMC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LAMC1. Alternate names: LAMB2, MGC87297ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-62266