Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMC1 Rat anti-Human, Mouse, Clone: SPM193, Abnova™
Rat monoclonal antibody raised against native mouse Lamc1.
Supplier: Abnova Corporation MAB13102
Specifications
LAMC1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
Lamb2 | |
Rat | |
Protein A/G purification | |
RUO | |
226519 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM193 | |
Rat monoclonal antibody raised against native mouse Lamc1. | |
In 10mM PBS. | |
Lamc1 | |
Murine EHS laminin preparation. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction