Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31138850UL

 View more versions of this product

Catalog No. NB121485


Add to cart

Description

Description

LAMC2 Polyclonal antibody specifically detects LAMC2 in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications

Specifications

LAMC2
Unconjugated
PBS, pH7.0
BM600, BM600-100kDa, Cell-scattering factor 140 kDa subunit, CSF, CSF 140 kDa subunit, EBR2, EBR2A, Epiligrin subunit gamma, kalinin (105kD), Kalinin subunit gamma, Kalinin/nicein/epiligrin 100 kDa subunit, kalinin-105kDa, ladsin (140kDa), Ladsin 140 kDa subunit, LAMB2Tlaminin, gamma 2 (nicein (100kD), kalinin (105kD), BM600 (100kD), Herlitzjunctional epidermolysis bullosa)), Laminin B2t chain, laminin subunit gamma-2, laminin, gamma 2, Laminin-5 subunit gamma, LAMNB2B2T, Large adhesive scatter factor 140 kDa subunit, MGC138491, MGC141938, nicein (100kDa), Nicein subunit gamma, nicein-100kDa
E. coli-derived Human LAMC2 fragment
Antigen and protein A Affinity-purified
RUO
Primary
3918
Western Blot, Immunofluorescence, Immunoprecipitation
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence -1:100-1:500, Immunoprecipitation -1-5-ul/mg
Purified
Rabbit
IgG
50 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit