Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen LAMC2
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence -1:100-1:500, Immunoprecipitation -1-5-ul/mg
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121484
View Documents
Novus Biologicals
NBP311388200UL
200 μg
Each for $320.71
Add to cart
 
NB121485
View Documents
Novus Biologicals
NBP31138850UL
50 μg
Each for $138.37
Add to cart
 
Description

Description

LAMC2 Polyclonal antibody specifically detects LAMC2 in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications

Specifications

LAMC2
Western Blot, Immunofluorescence, Immunoprecipitation
Purified
RUO
PBS, pH7.0
3918
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence -1:100-1:500, Immunoprecipitation -1-5-ul/mg
Unconjugated
Rabbit
Human
BM600, BM600-100kDa, Cell-scattering factor 140 kDa subunit, CSF, CSF 140 kDa subunit, EBR2, EBR2A, Epiligrin subunit gamma, kalinin (105kD), Kalinin subunit gamma, Kalinin/nicein/epiligrin 100 kDa subunit, kalinin-105kDa, ladsin (140kDa), Ladsin 140 kDa subunit, LAMB2Tlaminin, gamma 2 (nicein (100kD), kalinin (105kD), BM600 (100kD), Herlitzjunctional epidermolysis bullosa)), Laminin B2t chain, laminin subunit gamma-2, laminin, gamma 2, Laminin-5 subunit gamma, LAMNB2B2T, Large adhesive scatter factor 140 kDa subunit, MGC138491, MGC141938, nicein (100kDa), Nicein subunit gamma, nicein-100kDa
E. coli-derived Human LAMC2 fragment
Polyclonal
Antigen and protein A Affinity-purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit