Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A/C Monoclonal Antibody (JOL2), Biotin, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA546597

Encompass

Catalog No. PIMA546597


Add to Cart

Description

Description

Lamin A/C Monoclonal antibody specifically detects Lamin A/C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lamin A/C
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P02545
LMNA
Recombinant human lamin A
50 μg
Primary
Human
Liquid
Flow Cytometry, Immunocytochemistry, Western Blot
JOL2
Biotin
LMNA
70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg
Mouse
Protein G
RUO
4000
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG1
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit