Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A/C R453W Monoclonal Antibody (12A-2F5), Active Motif™
SDP

Mouse Monoclonal Antibody

Supplier:  Active Motif 39959

Encompass

 View more versions of this product

Catalog No. 50-199-3001


Add to Cart

Description

Description

Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).
Specifications

Specifications

Lamin A/C R453W
Monoclonal
1 mg/mL
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
P02545
LMNA
This Lamin A/C R453W antibody was raised against a peptide consisting of amino acids 449-457 of human Lamin A/C containing the R453W substitution.
100 μg
Primary
Human
Antibody
IgG1 κ
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
12A-2F5
Unconjugated
LMNA
70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A/C; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; Prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1
Mouse
Protein G
RUO
4000
-20°C, Avoid Freeze/Thaw Cycles
Liquid
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit