Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin A/C R482W Monoclonal Antibody (5H8-B4), Active Motif™

Mouse Monoclonal Antibody
$121.20 - $369.66
Specifications
Antigen | Lamin A/C R482W |
---|---|
Clone | 5H8-B4 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3006
![]() ![]() |
Active Motif
39964 |
10 μg |
Each for $121.20
|
|
50-199-3005
![]() ![]() |
Active Motif
39963 |
100 μg |
Each for $369.66
|
|
Description
Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).Specifications
Lamin A/C R482W | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
4000 | |
This Lamin A/C R482W antibody was raised against a recombinant protein corresponding to amino acids 430-545 of human Lamin A/C containing the R482W substitution. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
LMNA |
5H8-B4 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P02545 | |
LMNA | |
IgG1 | |
Protein G | |
Antibody |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title